Biophotometers, BioSpectrometer® kinetic
Catalog #
Product Details & Documents
The BioSpectrometer kinetic combines the specifications of the basic model and a heating element.
- Peltier element for temperature control of the shaft
- Preprogrammed and freely programmable kinetic methods
- Subsequent modification of the time window for regression evaluation of the kinetic curve
The cuvette shaft can be tempered in a range of 20 to 42 °C, which enables to carry out enzyme and substrate kinetics.