

Gram color staining kit, Sigma-Aldrich®
Catalog # 1.11885.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre carton
- Pk:5
- Seq:0005
- Pk Info:5× 500 ml (Preparation Capacity 250 slides)
Specifications
About this item
:
ADR:3,
:M-Clarity™ quality level = MQ300
UN:1993
:GHS02
:GHS07
Storage Temperature:Ambient
:1