Exact-Temp thermometers
Red or blue filling. Coating: PTFE/FEP, transparent.
- Available as digital or conventional thermometers (organic or mercury filling, with/without safety coating)
- Unbreakable plastic bottles, attached magnets for easy wall or door mounting, individual serial number
- All thermometers are encapsulated in a high grade safety coating to protect contents, users and environment from splintered glass or display fluid leakage (PTFE/FEP, transparent)
High precision instruments to monitor freezers, ultra-low freezers, blood bank freezers, refrigerators, incubators and ovens, as well as climate chambers.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Thermometers traceable to international standards (with certificate of conformance and accuracy).
Ordering information: Available with works certificate or with DAkkS calibration certificate (optional).
Delivery information: Supplied with certificate of conformity.
Red or blue filling. Coating: PTFE/FEP, transparent.
- Available as digital or conventional thermometers (organic or mercury filling, with/without safety coating)
- Unbreakable plastic bottles, attached magnets for easy wall or door mounting, individual serial number
- All thermometers are encapsulated in a high grade safety coating to protect contents, users and environment from splintered glass or display fluid leakage (PTFE/FEP, transparent)
High precision instruments to monitor freezers, ultra-low freezers, blood bank freezers, refrigerators, incubators and ovens, as well as climate chambers.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Certifications: Thermometers traceable to international standards (with certificate of conformance and accuracy).
Ordering information: Available with works certificate or with DAkkS calibration certificate (optional).
Delivery information: Supplied with certificate of conformity.
Documents
SCNL7000114_TDS_EN
Datasheet
Product Family Options
With safety coating
- DescriptionRangeDivisionLengthFillingImmersion depth
- Bottle thermometer for freezers−25...−5 °C0,1 °C235 mmBlueTotal1 * 1 SZT329 zł
- Bottle thermometer for refrigerators−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT263 zł
- Bottle thermometer for refrigerators, with DAkkS calibration certification−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT1 280 zł
- Bottle thermometer for blood bank freezers−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT339 zł
- Bottle thermometer for incubators+15...+30 °C0,1 °C180 mmBlueTotal1 * 1 SZT298 zł
- Bottle thermometer for incubators+25...+45 °C0,1 °C190 mmBlueTotal1 * 1 SZT305 zł
- Bottle thermometer for ovens+50...+115 °C0,5 °C200 mmBlueTotal1 * 1 SZT359 zł
- Bottle thermometer for freezers−90...+20 °C1,0 °C180 mmRedTotal1 * 1 SZT258 zł
- Bottle thermometer for freezers−30...0 °C0,5 °C130 mmRedTotal1 * 1 SZT202 zł
- Bottle thermometer for blood bank freezers−5...+20 °C0,5 °C150 mmRedTotal1 * 1 SZT261 zł
- Bottle thermometer for refrigerators−5...+15 °C0,5 °C130 mmRedTotal1 * 1 SZT187 zł
- Bottle thermometer for incubators+18...+50 °C0,5 °C130 mmRedTotal1 * 1 SZT194 zł
- Bottle thermometer for ovens+20...+130 °C1,0 °C135 mmRedTotal1 * 1 SZT213 zł
- Bottle thermometer for ovens+35...+200 °C1,0 °C180 mmRedTotal1 * 1 SZT611 zł
- Bottle thermometer for freezers−30...+0 °C0,5 °C115 mmMercuryTotal-
- Bottle thermometer for freezers−25...+5 °C0,1 °C210 mmMercuryTotal-
- Bottle thermometer for refrigerators−5...+15 °C0,5 °C125 mmMercuryTotal-
- Bottle thermometer for refrigerators−2...+10 °C0,1 °C180 mmMercuryTotal-
- Bottle thermometer for blood bank−2...+10 °C0,1 °C180 mmMercuryTotal-
- Bottle thermometer for incubators−18...+50 °C0,5 °C135 mmMercuryTotal-
- Bottle thermometer for incubators+15…+30 °C0,1 °C180 mmMercuryTotal-
- Bottle thermometer for ovens+20...+130 °C1,0 °C135 mmMercuryTotal-
- Bottle thermometer for incubators+25…+45 °C0,1 °C190 mmMercuryTotal-
- Bottle thermometer for ovens+35...+200 °C1,0 °C145 mmMercuryTotal-
- Bottle thermometer for ovens+95…+115 °C1,0 °C200 mmMercuryTotal-
Without safety coating
- DescriptionRangeDivisionLengthFillingImmersion depth
- Bottle thermometer for freezers−25...−5 °C0,1 °C235 mmBlueTotal1 * 1 SZT308 zł
- Bottle thermometer for freezers, with DAkkS calibration certification−25...−5 °C0,1 °C235 mmBlueTotal1 * 1 SZT1 360 zł
- Bottle thermometer for refrigerators−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT241 zł
- Bottle thermometer for refrigerators, with DAkkS calibration certification−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT1 250 zł
- Bottle thermometer for blood bank freezers−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT287 zł
- Bottle thermometer for blood bank freezers, with DAkkS calibration certification−2...+10 °C0,1 °C230 mmBlueTotal1 * 1 SZT1 320 zł
- Bottle thermometer for incubators+15...+30 °C0,1 °C180 mmBlueTotal1 * 1 SZT279 zł
- Bottle thermometer for incubators, with DAkkS calibration certification+15...+30 °C0,1 °C180 mmBlueTotal1 * 1 SZT1 310 zł
- Bottle thermometer for incubators+25...+45 °C0,1 °C190 mmBlueTotal1 * 1 SZT285 zł
- Bottle thermometer for incubators, with DAkkS calibration certification+25...+45 °C0,1 °C190 mmBlueTotal1 * 1 SZT1 320 zł
- Bottle thermometer for ovens+50...+115 °C0,5 °C240 mmBlueTotal1 * 1 SZT336 zł
- Bottle thermometer for ovens, with DAkkS calibration certification+50...+115 °C0,5 °C240 mmBlueTotal1 * 1 SZT1 410 zł
- Bottle thermometer for ultralow freezers−90...+20 °C1,0 °C180 mmRedTotal1 * 1 SZT241 zł
- Bottle thermometer for ultralow freezers, with DAkkS calibration certification−90...+20 °C1,0 °C180 mmRedTotal1 * 1 SZT1 250 zł
- Bottle thermometer for ultralow freezers−30...0 °C0,5 °C130 mmRedTotal1 * 1 SZT190 zł
- Bottle thermometer for ultralow freezers, with DAkkS calibration certification−30...0 °C0,5 °C130 mmRedTotal1 * 1 SZT1 160 zł
- Bottle thermometer for blood bank freezers−5...+20 °C0,5 °C150 mmRedTotal1 * 1 SZT247 zł
- Bottle thermometer for blood bank freezers, with DAkkS calibration certification−5...+20 °C0,5 °C150 mmRedTotal1 * 1 SZT1 260 zł
- Bottle thermometer for refrigerators−5...+15 °C0,5 °C130 mmRedTotal1 * 1 SZT175 zł
- Bottle thermometer for refrigerators, with DAkkS calibration certification−5...+15 °C0,5 °C130 mmRedTotal1 * 1 SZT1 130 zł
- Bottle thermometer for incubators+18...+50 °C0,5 °C130 mmRedTotal1 * 1 SZT181 zł
- Bottle thermometer for incubators, with DAkkS calibration certification+18...+50 °C0,5 °C130 mmRedTotal1 * 1 SZT1 140 zł
- Bottle thermometer for ovens+20...+130 °C1,0 °C135 mmRedTotal1 * 1 SZT200 zł
- Bottle thermometer for ovens, with DAkkS calibration certification+20...+130 °C1,0 °C135 mmRedTotal1 * 1 SZT1 180 zł
- Bottle thermometer for ovens+35...+200 °C1,0 °C180 mmRedTotal1 * 1 SZT593 zł