Disposable syringes, 3-parts, ONCE
Supplier: SEMADENI
Certificates
About this item
These syringes have precise pistons. The piston and seal are made of silicone rubber.
- Gas sterilised and non-pyrogenic
Sealing, sterility and depyrogenation are subject to continuous controls.
Certifications: Conforms with ISO 7886.
Packaging: Individually wrapped.
Frequently Bought Together