Przejdź do Nagłówek
Przejdź do Główna treść
Przejdź do Stopka
Zaawansowany
Zaawansowany
SDS
Certyfikaty
Dane producenta
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Microplate Seals
Adhesive PCR foil seal
Adhesive PCR foil seal
Adhesive PCR foil seal
Catalog # BZYM600228
Supplier:
BIOZYM
Expand image
Adhesive PCR foil seal
Catalog # BZYM600228
Supplier:
BIOZYM
Specifications
Description:
Adhesive PCR foil seal, sheets
L×W:
135×80 mm
Material:
Aluminium
Pk:
100
Specifications
About this item
Product Details
Product Details