GC syringes for Agilent instruments
Supplier: Thermo Fisher Scientific
Certificates
About this item
GC Autosampler syringes for use with Agilent GC instruments.
Available in a range of volumes from 5 to 10 µl.
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Syringes for gas chromatography
Supplier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Exhaust filters, V3.0
Supplier: S.C.A.T.
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals