Order Entry
Poland
ContactUsLinkComponent
Vinometer according to Bernadotte
Vinometer according to Bernadotte
Catalog # HECH44200010
undefined
Vinometer according to Bernadotte
Catalog # HECH44200010
Dane techniczne
  • Description:
    Vinometer
  • Material:
    Glass
  • Model:
    Bernadotte
  • Pk:
    1
  • Range:
    0 − 25
Dane techniczne
Informacje o tym produkcie
This instrument is designed for the accurate testing the percentage of alcohol in wine.