p-Anisic acid
Catalog # PROOERM-FC023
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:152.15 g/mol
Merck Index:13,00669
Boiling Pt:275 °C (1013 hPa)
Melting Pt:182…185 °C
Density:∼1.385 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-818-5
CAS Number:100-09-4
Formula:C₈H₈O₃
MDL Number:MFCD00002542