Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead

Lead

Health
Irritant
Environment
Supplier:   CHEMPUR
Lead
Supplier:  CHEMPUR
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 160 zł
    160 zł
    1 * 100 g
       160 zł

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 295 zł
    295 zł
    1 * 250 g
       295 zł

  • Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 463 zł
    463 zł
    1 * 500 g
       463 zł