Chlorotrimethylsilane 1 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL384410-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
:108.64 g/mol
EINECS:200-900-5
CAS Number:75-77-4
Formula:C₃H₉ClSi
MDL Number:MFCD00000502