Iodine 0.05 mol concentrated aqueous solution
Catalog # POCH491265165-1EA
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Environmentally Preferable:
Specifications
About this item
:
:253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
CAS Number:7553-56-2
Formula:I₂
MDL Number:MFCD00011355