
Tetrachloropicolinic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10469-09-7
:
Formula:C₆HCl₄NO₂
MDL Number:MFCD00185833
:260.89 g/mol
Melting Pt:171…173 °C
EINECS:233-956-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together