4-Sulphobenzoic acid monopotassium salt
Catalog # MOLE65542831-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:240.28 g/mol
EINECS:226-435-8
CAS Number:5399-63-3
Formula:C₇H₅KO₅S
MDL Number:MFCD00007509