Order Entry
Poland
ContactUsLinkComponent
Lead(II) methanesulphonate 50% in aqueous solution, Sigma-Aldrich®
Lead(II) methanesulphonate 50% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL462667-1L
Supplier:  Merck
CAS Number:  
Lead(II) methanesulphonate 50% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL462667-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:401-750-5
CAS Number:17570-76-2
Formula:C₂H₆O₆PbS₂
MDL Number:MFCD00137736