Order Entry
Poland
ContactUsLinkComponent
1,3-Butadiene 20% in toluene, Sigma-Aldrich®
1,3-Butadiene 20% in toluene, Sigma-Aldrich®
Catalog # SIAL695580-250G
Supplier:  Merck
CAS Number:  
1,3-Butadiene 20% in toluene, Sigma-Aldrich®
Catalog # SIAL695580-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :54.09 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:203-450-8
CAS Number:106-99-0
Formula:C₄H₆
MDL Number:MFCD00008659