Order Entry
Poland
ContactUsLinkComponent
2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine ≥97%
Irritant
2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine ≥97%
Catalog # B25741.03
CAS Number:  
Irritant
2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine ≥97%
Catalog # B25741.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:93438-27-8
 :
Formula:C₁₇H₂₂N₆O₂
MDL Number:MFCD00272422
Melting Pt:171…174 °C
 :342.40 g/mol
 :GHS07
Storage Temperature:Ambient