4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:19438-60-9
:
Formula:C₉H₁₂O₃
MDL Number:MFCD00005927
:M-Clarity™ quality level = MQ100
:168.19 g/mol
EINECS:243-072-0
Frequently Bought Together