Eosin Y (yellowish) 1% in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:17372-87-1
:
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040
:691.86 g/mol
Storage Temperature:Ambient
EINECS:241-409-6
Frequently Bought Together