1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole
Catalog # MOLE37688754-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:194.21 g/mol
EINECS:257-909-2
CAS Number:52431-78-4
Formula:C₇H₆N₄OS
MDL Number:MFCD00132898