Hanus solution (Iodine bromine) c(IBr) = 0.1 mol/l, Titripur® for determination of iodine value, Supelco®
Supplier: Merck
Certificates
About this item
:7789-33-5
Formula:IBr
MDL Number:MFCD00011353
Density:1.06 g/cm3 (20 °C)
ADR:8 (3),II
:M-Clarity™ quality level = MQ100
Flash Pt:40 °C
UN:2789
:GHS02
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Smart Chemical Products
Product Literature
msds_109164
SDS
Frequently Bought Together