Pallets and sumps for KTC, WSP
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
These pallets and sumps made of 3 mm sheet steel are ideal for storing and filling off hazardous substances from large containers.
- Sump capacity of 1000 L
- Support surface is a removable fire-galvanised grid
- Ground clearance 100 mm for fork-lift truck transport
- High corrosion resistance – with primer and painting or fire galvanisation
Frequently Bought Together