



n-Pentane for HPLC
Catalog # CHMP617179209.2500
Supplier: CHEMPUR
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
Specifications
About this item
:
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498