
Methyl red 1 g/l in ethanol
: CHEMPUR
:493-52-7
:
:C₁₅H₁₅N₃O₂
:MFCD00002425
:3,II
:269.3 g/mol
:207-776-1
:1170
:GHS02
:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals