
Tetramethylammonium acetate hydrate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1266615-86-4
:
Formula:C₆H₁₅NO₂·xH₂O
MDL Number:MFCD03066126
:133.20 g/mol (anhy)
EINECS:234-179-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline solid
- Infrared spectrum:Conforms
- Water:≤14 % (K.F.)
- Titration with HClO4:≥94 % (On dry substance)
Specification Test Results
Frequently Bought Together