(R)-(-)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:39648-67-4
:
Formula:C₂₀H₁₃O₄P
MDL Number:MFCD00010045
:348.289 g/mol
:M-Clarity™ quality level = MQ100
Frequently Bought Together