Order Entry
Poland
ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 10% in aqueous solution, Reagent Grade
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 10% in aqueous solution, Reagent Grade
Catalog # N907-1L
Supplier:  VWR Chemicals
CAS Number:  
undefined
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 10% in aqueous solution, Reagent Grade
Catalog # N907-1L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
 :
 :614.89 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083