Order Entry
Poland
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) 0.02% in aqueous solution, Sigma-Aldrich®
EDTA (ethylenediamine tetraacetic acid) 0.02% in aqueous solution, Sigma-Aldrich®
Catalog # SIALE8008-100ML
Supplier:  Merck
CAS Number:  
undefined
EDTA (ethylenediamine tetraacetic acid) 0.02% in aqueous solution, Sigma-Aldrich®
Catalog # SIALE8008-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :292.25 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541