Sodium hydroxide 0.5 mol/l (0.5 N) in methanol
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:3286
ADR:3,II
Frequently Bought Together