
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.02 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
:GHS07
Frequently Bought Together