Ammonium sulphate 0.1 mol/l in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7783-20-2
:
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
:132.14 g/mol
Storage Temperature:Ambient
EINECS:231-984-1
Frequently Bought Together