Sodium hydroxide 0.01 mol concentrated aqueous solution
1
Catalog # CHMP168109338.0001
Supplier: CHEMPUR
Sodium hydroxide 0.01 mol concentrated aqueous solution
Catalog # CHMP168109338.0001
Supplier: CHEMPUR
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:∼105 °C (1013 hPa)
- Density:1.327 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Specifications
Specifications
- Pk:1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



