0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Sodium hydroxide
Sodium hydroxide 1 mol concentrated aqueous solution
Sodium hydroxide 1 mol concentrated aqueous solution
CHMP168109337.0001
:
CHEMPUR
:
Sodium hydroxide 1 mol concentrated aqueous solution
CHMP168109337.0001
:
CHEMPUR
:
Pk:
1
Seq:
0001
:
:
40 g/mol
:
∼105 °C (1013 hPa)
:
1.327 g/cm³ (20 °C)
:
Ambient
:
215-185-5
:
1824
:
8,II
:
GHS05
:
1310-73-2
:
NaOH
:
MFCD00003548