Order Entry
Poland
ContactUsLinkComponent
Hydrogen peroxide 35%
  CHMP428851221.1000
 :  CHEMPUR
 :  
Hydrogen peroxide 35%
  CHMP428851221.1000
 :  CHEMPUR
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
 :
 :∼107 °C (1013 hPa)
 :< 0 °C
 :1.11 g/cm³ (20 °C)
 :5.1,II
 :34.01 g/mol
 :231-765-0
 :2014
 :GHS05
 :GHS07
 :Ambient
 :7722-84-1
 :H₂O₂
 :MFCD00011333