Hydrogen peroxide 3,5% USP, J.T.Baker®
4 L
Catalog # 2182-03
Supplier: Avantor
Jump to:
Product Details & Documents
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:100 °C (1013 hPa)
- Density:1.01 g/cm³ (20 °C)
- :Density: 1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- Identification:Passes Test
- Acidity:Passes Test
- Nonvolatile Residue (USP),mg.:≤30
- Barium (Ba):Passes Test
- Limit of Preservative:≤0.05 %
- Assay (g/100mL):2.5 - 3.5
- Particulate Matter:Passes Test
Specifications
- Pk:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



