Order Entry
Poland
ContactUsLinkComponent
Nitric acid 55%
Corrosive
Nitric acid 55%
Catalog # CHMP325296030.5000
Supplier:  CHEMPUR
CAS Number:  
Corrosive
Nitric acid 55%
Catalog # CHMP325296030.5000
Supplier:  CHEMPUR
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349