
Potassium hydroxide 40% in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
:GHS05
Frequently Bought Together