0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Potassium hydroxide
Potassium hydroxide 0.5 mol in aqueous solution
Potassium hydroxide 0.5 mol in aqueous solution
CHMP167468239.0001
:
CHEMPUR
:
Potassium hydroxide 0.5 mol in aqueous solution
CHMP167468239.0001
:
CHEMPUR
:
Pk:
1
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
1814
:
8,II
:
GHS05
:
1310-58-3
:
KOH
:
MFCD00003553