Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide 1 mol in aqueous solution
Potassium hydroxide 1 mol in aqueous solution
CHMP167468237.0001
:
CHEMPUR
:
Potassium hydroxide 1 mol in aqueous solution
CHMP167468237.0001
:
CHEMPUR
:
Pk:
1
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
1814
:
8,II
:
GHS05
:
1310-58-3
:
KOH
:
MFCD00003553