Ammonium thiocyanate 1 mol/l (1 N) in aqueous solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:1762-95-4
:
Formula:NH₄SCN
MDL Number:MFCD00011428
:76.12 g/mol
Storage Temperature:Ambient
EINECS:217-175-6
Frequently Bought Together