Methyl orange 0,04% in aqueous solution
Supplier: CHEMPUR
Jump to:
About this item
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
- CAS Number:547-58-0
- Formula:C₁₄H₁₄N₃NaO₃S
- MDL Number:MFCD00007502
- Molecular Weight:327.34 g/mol
- Storage Temperature:Ambient
- EINECS:208-925-3
4 Options Available Below
Product Details & Documents
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
- CAS Number:547-58-0
- Formula:C₁₄H₁₄N₃NaO₃S
- MDL Number:MFCD00007502
- Molecular Weight:327.34 g/mol
- Storage Temperature:Ambient
- EINECS:208-925-3
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



