
3-Hexadecylthiophene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:119269-24-8
:
Formula:C₂₀H₃₆S
MDL Number:MFCD07368973
:308.57 g/mol
Melting Pt:27…32 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together