Order Entry
Poland
ContactUsLinkComponent
4,5-Difluoro-2-hydroxybenzonitrile ≥99%
Toxic
Irritant
4,5-Difluoro-2-hydroxybenzonitrile ≥99%
Catalog # H26155.03
CAS Number:  
Toxic
Irritant
4,5-Difluoro-2-hydroxybenzonitrile ≥99%
Catalog # H26155.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

CAS Number:186590-36-3
 :
Formula:C₇H₃F₂NO
MDL Number:MFCD08275287
 :155.10 g/mol
Melting Pt:161…163 °C
EINECS:None
ADR:6.1,III
 :GHS06
 :GHS07
Storage Temperature:Ambient