Methyl orange 0,1% in ethanol
Supplier: CHEMPUR
Certificates
About this item
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
CAS Number:547-58-0
:
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502
Boiling Pt:90
Molecular Weight:327.34 g/mol
Flash Pt:38
ADR:3 III
Merck Index:13,06127
EINECS:208-925-3
UN:1170
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together