Boron trichloride 1 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10294-34-5
:
Formula:BCl₃
MDL Number:MFCD00011313
:117.17 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:233-658-4
Frequently Bought Together