Order Entry
Poland
ContactUsLinkComponent
5-(Methylthio)-1H-tetrazole
5-(Methylthio)-1H-tetrazole
Catalog # MOLE14815228-5G
Supplier:  Molekula
CAS Number:  
5-(Methylthio)-1H-tetrazole
Catalog # MOLE14815228-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
CAS Number:29515-99-9
Formula:C₂H₄N₄S
MDL Number:MFCD00068726