Order Entry
Poland
ContactUsLinkComponent
Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution, Sigma-Aldrich®
Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL401579-250ML
Supplier:  Merck
CAS Number:  
Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution, Sigma-Aldrich®
Catalog # SIAL401579-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :380.81 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:237-486-0
CAS Number:13814-96-5
Formula:B₂F₈Pb
MDL Number:MFCD00016270