Nitronium tetrafluoroborate 0.5 mol/l in sulpholane, Sigma-Aldrich®
Catalog # SIAL176303-200ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:200 mL
- Seq:2530
Specifications
About this item
:
:132.81 g/mol
:M-Clarity™ quality level = MQ100
EINECS:237-533-5
CAS Number:13826-86-3
Formula:BF₄NO₂
MDL Number:MFCD00011432