Order Entry
Poland
ContactUsLinkComponent
Nitronium tetrafluoroborate 0.5 M in sulpholane
Irritant
Corrosive
Nitronium tetrafluoroborate 0.5 M in sulpholane
Certificates
undefined
Irritant
Corrosive
Nitronium tetrafluoroborate 0.5 M in sulpholane

Specification Test Results

  • Appearance:
    Colorless to yellow Solution
  • Strength:
    0.48 to 0.55 M
  • Melting point:
    88 to 92 °C
  • Loss on drying:
    ≤0.5 % (1 g, 60 °C, 4 h)
  • HPLC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together