Order Entry
Poland
ContactUsLinkComponent
Ammonium sulphate-[¹⁵N₂] (60% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂] (60% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488097-5G
Supplier:  Merck
CAS Number:  
Ammonium sulphate-[¹⁵N₂] (60% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL488097-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:43086-58-4
Formula:H₈N₂O₄S
MDL Number:MFCD00003392