Skip to Header
Skip to Main Content
Skip to Footer
Zaawansowany
Zaawansowany
SDS
Certyfikaty
Dane producenta
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate-[15N2]
Ammonium sulphate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
Ammonium sulphate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
SIAL488089-250G
:
Merck
:
Ammonium sulphate-[¹⁵N₂] (5% ¹⁵N), Sigma-Aldrich®
SIAL488089-250G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
250 g
Seq:
3545
:
:
M-Clarity™ quality level = MQ200
:
Ambient
:
43086-58-4
:
H₈N₂O₄S
:
MFCD00003392